Sodium Molybdate 2-hydrate (Reag. Ph. Eur.) for analysis, ACS 1 * 1.000 g
Catalog # APPC131701.1211
Supplier: PanReac AppliChem
Sodium Molybdate 2-hydrate (Reag. Ph. Eur.) for analysis, ACS 1 * 1.000 g
Catalog # APPC131701.1211
Supplier: PanReac AppliChem
Frequently Bought Together
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Methanol ≥99.8% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Hexane (mixture of isomers) 95+% for spectroscopy
Supplier: Thermo Fisher Scientific
1,3-Propanediol ≥99%
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals