di-Sodium Tartrate 2-hydrate standard for volumetry, ACS 1 * 100 g
Catalog # APPC241719.1608
Supplier: PanReac AppliChem
di-Sodium Tartrate 2-hydrate standard for volumetry, ACS 1 * 100 g
Catalog # APPC241719.1608
Supplier: PanReac AppliChem
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals