Order Entry
Austria
Orders LinkContactUsLinkComponent
qScript™ One-Step qRT-PCR kits

qScript™ One-Step qRT-PCR kits

qScript™ One-Step qRT PCR Kit, with ROX, Agilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480, 200 Assays
Catalog # 733-1259
Supplier:   Quantabio
qScript™ One-Step qRT-PCR kits
Catalog # 733-1259
Supplier:  Quantabio
CAS Number:  
qScript™ One-Step qRT-PCR kits
qScript™ One-Step qRT-PCR kits

qScript™ One-Step qRT PCR Kit, with ROX, Agilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480, 200 Assays

CAS Number:  
Price
687,00 €
Log in for availability
Product Details & Documents

Synthesis of cDNA and PCR amplification are carried out in the same tube without opening between procedures. These kits are optimised to deliver maximum RT-PCR efficiency and specificity, enabling unbiased co-amplification of low copy transcripts in the presence of higher copy reference genes. As a result the qScript™ One-Step qRT-PCR Kits offer improved sensitivity over other commercially available qPCR reagents. The qScript™ One-Step Master Mix maximises the activities of both reverse transcriptase and PCR enzymes, while minimising primer-dimer formation. High specificity is assured with AccuStart™ Taq DNA Polymerase, which is bound by monoclonal antibodies for superior hot-start. Upon heating to 95 °C, the antibodies denature irreversibly releasing fully active and unmodified Taq DNA polymerase. qScript™ One-Step qRT-PCR Kits are available for all Real-Time PCR instrument platforms, including those requiring normalisation with ROX reference dye.

  • Broad dynamic range ensures reliable data
  • Highly sensitive for low copy RNA quantification
  • Unbiased duplex amplification for more accurate results
  • Simple, fast protocol with less variability

Documents

Specifications

Specifications

  • Cycler compatibility:
    Agilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480
  • Pk:
    200 Assays
  • Description:
    qScript™ One-Step qRT PCR Kit, with ROX
  • Cat. No.:
    733-1259
  • One step:
    Yes
  • Application:
    RT-qPCR probe

Specifications