PREDNICARBATE FOR SYSTEM SUITABILITY 1 * 10 mg
Catalog # EDQMY0001630
Supplier: EDQM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
BUFFER PH 9,21 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Conductivity standards
Supplier: HAMILTON BONADUZ
Ammonia solution 6 mol/l (6 N) Reag. Ph. Eur. 1004702
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, pH electrode maintenance kit
Supplier: VWR Chemicals
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Ethanolamine, Supelco®
Supplier: Merck
Benzophenone, Supelco®
Supplier: Merck
Supelco® Organic Reference Standard, Scopoletin
Supplier: Merck