Order Entry
Austria
Orders LinkContactUsLinkComponent
Squared bossheads

Squared bossheads

Stainless steel 18/10, 12 - 13 × 12 - 13 mm
Catalog # BOCHB08599125
Supplier:   BOCHEM
Squared bossheads
Catalog # BOCHB08599125
Supplier:  BOCHEM
Squared bossheads
Squared bossheads

Stainless steel 18/10, 12 - 13 × 12 - 13 mm

Price
42,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Squared bossheads with hexagon socket screws.

  • Made from 18/10 stainless steel or aluminium (laboral)
  • Angle: 90°
Specifications
  • Material:
    Stainless steel 18/10
  • For rod Ø:
    12 - 13 × 12 - 13 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.