Orders LinkContactUsLinkComponent
Dropping pipettes
Dropping pipettes
Dropping pipettes with silicone teat, for 50 ml bottles, joint size 14/15
Catalog # BIBBSBD050
Supplier:   SciLabware
Dropping pipettes
Catalog # BIBBSBD050
Supplier:  SciLabware
Dropping pipettes
Dropping pipettes
Dropping pipettes with silicone teat, for 50 ml bottles, joint size 14/15
Price
85,60 €
Log in for availability
1 * 5 ST
Product Details & Documents
Soda-lime glass.
  • With integral ground glass cone on pipette
  • Supplied with rubber teats
  • Bottles available separately
Specifications
  • Description:
    Dropping pipettes with silicone teat, for 50 ml bottles, joint size 14/15
  • Seq:
    01