Orders LinkContactUsLinkComponent
Preprinted labels
Preprinted labels
Colour-coded dot labels, Paper, Red
Catalog # EPSC503-0005
   Low Manufacturing Impact
Supplier:   EP SCIENTIFIC
Preprinted labels
Catalog # EPSC503-0005
Supplier:  EP SCIENTIFIC
   Low Manufacturing Impact
Preprinted labels
Preprinted labels
Colour-coded dot labels, Paper, Red
Price
234,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Label is colour-coded with the chemical preservative type for quick reference and safe handling. Sticks securely to plastic or glass sample containers. Provides easy sample aliquot identification and enhances safe handling information visibility.
Specifications
  • Colour:
    Red
  • Description:
    Colour-coded dot labels
  • Material:
    Paper
  • Seq:
    02
  • Cat. No.:
    EPSC503-0005
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact