Order Entry
Austria
Orders LinkContactUsLinkComponent
Disposable filters, Optimizer® DEV and DI

Disposable filters, Optimizer® DEV and DI

DEV-L disposable filter with 1/4" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×175 mm, 0,05 µm, 3600 cm²
Catalog # ENTECHUZ0L0P1
Supplier:   ENTEGRIS
Disposable filters, Optimizer® DEV and DI
Catalog # ENTECHUZ0L0P1
Supplier:  ENTEGRIS
Disposable filters, Optimizer® DEV and DI
Disposable filters, Optimizer® DEV and DI

DEV-L disposable filter with 1/4" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×175 mm, 0,05 µm, 3600 cm²

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

These point-of-use filters meet small to mid-volume photochemical filtration needs for positive developer and DI water applications. Both Optimizer® DEV filters (for developer applications) and Optimizer® DI filters (for DI water applications) require no pre-wetting, so the filters function immediately after installation. The design also limits the handling of hazardous chemicals during the installation and promotes a healthier workplace. Do not use either filter with aromatic hydrocarbons.

  • Manufactured, tested, and packaged in a world-class, ISO 9001 Quality Systems Standard facility
  • O-ringless connections avoid potential sources of contamination

Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UHMWPE); Supports, core and sleeve: HDPE; Disposable shell: PP

Maximum operating temperature: 60 °C

Maximum forward differential pressure: 2,8 bar at 25 °C

Maximum reverse differential pressure: 2,8 bar at 25 °C

Articles may not be available in all countries. Please contact your local Avantor office.

Specifications
  • Filtration area:
    3600 cm²
  • Pore size:
    0,05 µm
  • Type:
    DEV-L disposable filter with 1/4" connection, 1/4" vent/drain Super Type Pillar® fittings, Ø×L: 71×175 mm
  • Seq:
    15