Order Entry
Austria
Orders LinkContactUsLinkComponent
Disposable filters, Optimizer® DEV and DI | Avantor

Disposable filters, Optimizer® DEV and DI

DI-L disposable filter with 8 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×205 mm, 0,2 µm, 3600 cm²
Artikelnr. ENTECBUG8L0P1
Anbieter:   ENTEGRIS
Disposable filters, Optimizer® DEV and DI
Artikelnr. ENTECBUG8L0P1
Anbieter:  ENTEGRIS
Disposable filters, Optimizer® DEV and DI
Disposable filters, Optimizer® DEV and DI

DI-L disposable filter with 8 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×205 mm, 0,2 µm, 3600 cm²

Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

These point-of-use filters meet small to mid-volume photochemical filtration needs for positive developer and DI water applications. Both Optimizer® DEV filters (for developer applications) and Optimizer® DI filters (for DI water applications) require no pre-wetting, so the filters function immediately after installation. The design also limits the handling of hazardous chemicals during the installation and promotes a healthier workplace. Do not use either filter with aromatic hydrocarbons.

  • Manufactured, tested, and packaged in a world-class, ISO 9001 Quality Systems Standard facility
  • O-ringless connections avoid potential sources of contamination

Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UHMWPE); Supports, core and sleeve: HDPE; Disposable shell: PP

Maximum operating temperature: 60 °C

Maximum forward differential pressure: 2,8 bar at 25 °C

Maximum reverse differential pressure: 2,8 bar at 25 °C

Articles may not be available in all countries. Please contact your local Avantor office.

Spezifikationen
  • Filtration area:
    3600 cm²
  • Pore size:
    0,2 µm
  • Type:
    DI-L disposable filter with 8 mm connections, 4 mm vent/drain Super Type Pillar® fittings, Ø×L: 71×205 mm
  • Seq:
    58
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.