Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Standard reference materials and buffer solutions, CertiPUR®, Supelco®

Sodium hydrogen carbonate/sodium carbonate, 10,014 (25°C), Glass bottle for solids, 2×25 g, 2
Catalog # 1.01962.0001
Supplier:   Merck
Standard reference materials and buffer solutions, CertiPUR®, Supelco®
Catalog # 1.01962.0001
Supplier:  Merck
CAS Number:  
Standard reference materials and buffer solutions, CertiPUR®, Supelco®

Sodium hydrogen carbonate/sodium carbonate, 10,014 (25°C), Glass bottle for solids, 2×25 g, 2

CAS Number:  
Price
367,00 €
Log in for availability
1 * 2 ST
Product Details & Documents

Materials for the precise calibration and monitoring of pH instruments.

  • Traceable to NIST primary standard and PTB
  • Maximum deviation ±0,003 pH units
  • Measured in an independent accredited calibration laboratory for the pH value measurement (DKD-K-14301)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2
  • Description:
    Sodium hydrogen carbonate/sodium carbonate
  • Pk Info:
    2×25 g
  • pH value:
    10,014 (25°C)
  • Cat. No.:
    1.01962.0001

Specifications