Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Sodium chloride standard solution, pour-top, 491 mg/l NaCl, 1000 µS/cm, 50 mL
Catalog # HACH1440026
Supplier:   Hach
Conductivity standards
Catalog # HACH1440026
Supplier:  Hach
CAS Number:  
Conductivity standards
Conductivity standards
Sodium chloride standard solution, pour-top, 491 mg/l NaCl, 1000 µS/cm, 50 mL
CAS Number:  
Price
16,60 €
Log in for availability
1 * 50 mL
Product Details & Documents
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Specifications
Specifications
  • Pk:
    50 mL
  • Description:
    Sodium chloride standard solution, pour-top, 491 mg/l NaCl
  • Conductivity:
    1000 µS/cm