ContactUsLinkComponent
C18H x 5µm particle size x 120Å pore size, 10mm length x 300µm ID, PEEKsil ProteCol HPLC Trap Column 1 * 1 items
Catalog # SGEA2C182-05T30K
C18H x 5µm particle size x 120Å pore size, 10mm length x 300µm ID, PEEKsil ProteCol HPLC Trap Column 1 * 1 items
Catalog # SGEA2C182-05T30K
Frequently Bought Together