ContactUsLinkComponent
XT STARTER M-PCR system for ice-free PCR/QPCR preparation. Low profile, open platform ice-free cooler holds 12 x 1,5 mL or 2,0 mL microfuge tubes and 6 rows of PCR stripwells. 1 * 1 items
Catalog # BCISBCS-557
Supplier:  BIOCISION
undefined
XT STARTER M-PCR system for ice-free PCR/QPCR preparation. Low profile, open platform ice-free cooler holds 12 x 1,5 mL or 2,0 mL microfuge tubes and 6 rows of PCR stripwells. 1 * 1 items
Catalog # BCISBCS-557
Supplier:  BIOCISION
Frequently Bought Together