ContactUsLinkComponent
Chromosorb PAW 12% UC W982, 80/100, preconditioned, 41.28 mm coil, 2 feet (0.61 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Catalog # HEWLG3591-81200
Supplier:  Agilent
Chromosorb PAW 12% UC W982, 80/100, preconditioned, 41.28 mm coil, 2 feet (0.61 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Catalog # HEWLG3591-81200
Supplier:  Agilent
Frequently Bought Together