Precision hydrometers, ASTM E100
138 H-62, 1,650 - 1,700 sp/gr, 0,001 sp/gr, 260 mm
SCNL3222024
These precision ASTM E100 hydrometers are designed for specific gravity or relative density.
- Reference temperature: 15,5 °C (60/60 °F)
- Without thermometer
- Suitable for calibration
: Available with either a works or a DAkkS calibration certificate on request
: Each packed in a square plastic sleeve.
- Length:260 mm
- Seq:21
- Division:0,001 sp/gr
- Model:138 H-62
- Range:1,650 - 1,700 sp/gr
- Cat. No.:SCNL3222024
- Reference temperature:15,5 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



