Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Inserts

E 142 Insert for different instruments, equipped with 2 hinged handles, 1 mm wire, 5 mm mesh size, 5 mm frame (255×480×55 mm), PG 8581, PG 8591, 225×480×55 mm
Catalog # MIEL3830300
Supplier:   MIELE
Inserts
Catalog # MIEL3830300
Supplier:  MIELE
Inserts
Inserts

E 142 Insert for different instruments, equipped with 2 hinged handles, 1 mm wire, 5 mm mesh size, 5 mm frame (255×480×55 mm), PG 8581, PG 8591, 225×480×55 mm

Price
198,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 142 Insert for different instruments, equipped with 2 hinged handles, 1 mm wire, 5 mm mesh size, 5 mm frame (255×480×55 mm)
  • Seq:
    6
  • W×D×H:
    225×480×55 mm
  • For:
    PG 8581, PG 8591
  • Plumbing:
    1× cold water (CW);
    1× cold water for steam condenser;
    1× de-mineralised water (AD);
    1× hot water (HW);
    1 drain hose connection
  • Cat. No.:
    MIEL3830300
  • Heater:
    5.4 kW
  • Dispensers:
    184 dispenser pump for liquid detergent;
    1 dispenser pump for neutralising agent