Orders LinkContactUsLinkComponent
FlashGel® camera
FlashGel® camera
FlashGel® device pack, includes camera, dock and a power supply
Catalog # LONZ57062
Supplier:   LONZA
FlashGel® camera
Catalog # LONZ57062
Supplier:  LONZA
FlashGel® camera
FlashGel® camera
FlashGel® device pack, includes camera, dock and a power supply
Price
3 180,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
The FlashGel® camera captures data from the FlashGel® system, allowing complete separation, recovery and documentation safely, at the bench, in minutes. This simple digital camera in an enclosed hood connects directly to a laptop, PC or PDA via USB.
  • Complete gel run and image capture in five minutes
  • Photograph gels at the bench, without UV light
  • Sharp, clear, high-resolution images
  • Simple user interface
  • Optimised exposure for FlashGel® cassettes
Hood (W×L×H): 100×110×160 mm
Image file type: .JPG, .TIFF, .PDF
Specifications
  • Description:
    FlashGel® device pack, includes camera, dock and a power supply
  • Seq:
    02