ContactUsLinkComponent
BIOSENSOR / ANTI-HUMAN FC CAPTURE (AHC) - Pack of 5 trays of 96 biosensors 1 * 5 items
Catalog # PALF18-5063
Supplier:  PALL FORTEBIO
BIOSENSOR / ANTI-HUMAN FC CAPTURE (AHC) - Pack of 5 trays of 96 biosensors 1 * 5 items
Catalog # PALF18-5063
Supplier:  PALL FORTEBIO
Frequently Bought Together