ContactUsLinkComponent
qPCR detection of Clostridium difficile (toxin B). Contains: Primer and probe mix, Reverse Transcription target specific primers (RN A genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-C.DIB
Supplier:  PRIMERDESIGN
qPCR detection of Clostridium difficile (toxin B). Contains: Primer and probe mix, Reverse Transcription target specific primers (RN A genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-C.DIB
Supplier:  PRIMERDESIGN
Frequently Bought Together