qPCR detection of Tsukamurella inchonensis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curv e, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-T.INCHONE
Supplier: PRIMERDESIGN
Frequently Bought Together