Ref gene selection kit dog. 12 most cited genes (primer only) plus qbase+ licence 1 * 1 items
GE-SY-12-CF
Ref gene selection kit dog. 12 most cited genes (primer only) plus qbase+ licence 1 * 1 items
GE-SY-12-CF
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection