ContactUsLinkComponent
qPCR detection of Mycoplasma haemofelis. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome v iruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-M.HAE
Supplier:  PRIMERDESIGN
qPCR detection of Mycoplasma haemofelis. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome v iruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-M.HAE
Supplier:  PRIMERDESIGN
Frequently Bought Together