ContactUsLinkComponent
qPCR detection of Avian Influenza A Virus Subtype H5. Contains: Primer and probe, RT target specific primers (RNA viruses only), Sta ndard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-H5
Supplier:  PRIMERDESIGN
qPCR detection of Avian Influenza A Virus Subtype H5. Contains: Primer and probe, RT target specific primers (RNA viruses only), Sta ndard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-H5
Supplier:  PRIMERDESIGN
Frequently Bought Together