ContactUsLinkComponent
qPCR detection of Streptococcus salivarius. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genom e viruses only), Copy number standard curve 1 * 150 Tests
Artikelnr. PATH-STD-S.SAL
Lieferant:  PRIMERDESIGN
qPCR detection of Streptococcus salivarius. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genom e viruses only), Copy number standard curve 1 * 150 Tests
Artikelnr. PATH-STD-S.SAL
Lieferant:  PRIMERDESIGN
Frequently Bought Together