ContactUsLinkComponent
qPCR detection of all Neisseria meningitidis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard cu rve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-N.MENINGI
Supplier:  PRIMERDESIGN
qPCR detection of all Neisseria meningitidis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard cu rve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-N.MENINGI
Supplier:  PRIMERDESIGN
Frequently Bought Together