ContactUsLinkComponent
qPCR detection of Rift Valley Fever Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-RVFV-
Supplier:  PRIMERDESIGN
qPCR detection of Rift Valley Fever Virus. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-RVFV-
Supplier:  PRIMERDESIGN
Frequently Bought Together