ContactUsLinkComponent
qPCR detection of Methicillin-resistant Staphylococcus aureus with Staphylococcal cassette chromosome mec(SCC mec) type IVa. Contain s: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction co 1 * 150 Tests
Catalog # PATH-MRSA-SCC4
Supplier:  PRIMERDESIGN
qPCR detection of Methicillin-resistant Staphylococcus aureus with Staphylococcal cassette chromosome mec(SCC mec) type IVa. Contain s: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction co 1 * 150 Tests
Catalog # PATH-MRSA-SCC4
Supplier:  PRIMERDESIGN
Frequently Bought Together