ContactUsLinkComponent
qPCR detection of Bordetella Bronchiseptica and Bordetella Parapertussis. Contains: Primer and probe, RT target specific primers (RN A viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-B.BRONCHI
Supplier:  PRIMERDESIGN
qPCR detection of Bordetella Bronchiseptica and Bordetella Parapertussis. Contains: Primer and probe, RT target specific primers (RN A viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-B.BRONCHI
Supplier:  PRIMERDESIGN
Frequently Bought Together