ContactUsLinkComponent
qPCR detection of Human Immunodeficiency Virus type 1. Contains: Primer and probe, RT target specific primers (RNA viruses only), St andard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-HIV1
Supplier:  PRIMERDESIGN
qPCR detection of Human Immunodeficiency Virus type 1. Contains: Primer and probe, RT target specific primers (RNA viruses only), St andard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # Z-PATH-HIV1
Supplier:  PRIMERDESIGN
Frequently Bought Together