ContactUsLinkComponent
qPCR detection of Human Parainfluenza virus type 4B. Contains: Primer and probe, RT target specific primers (RNA viruses only), Stan dard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HPIV4B
Supplier:  PRIMERDESIGN
qPCR detection of Human Parainfluenza virus type 4B. Contains: Primer and probe, RT target specific primers (RNA viruses only), Stan dard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-HPIV4B
Supplier:  PRIMERDESIGN
Frequently Bought Together