ContactUsLinkComponent
CLEANROOM STAINLESS STEEL HEIGHT ADJUSTABLE TABLES WITH CUT OUT TABLE TOP DESIGN 1200x860x767-1147mm MAX LOAD 100KG ELECTRICAL TABLE HEIGHT ADJUSTMENT SYSTEM WITH MEMORY FUNCTION 1 * 1 items
Catalog # PALBPCWU1299
Supplier:  Palbam
CLEANROOM STAINLESS STEEL HEIGHT ADJUSTABLE TABLES WITH CUT OUT TABLE TOP DESIGN 1200x860x767-1147mm MAX LOAD 100KG ELECTRICAL TABLE HEIGHT ADJUSTMENT SYSTEM WITH MEMORY FUNCTION 1 * 1 items
Catalog # PALBPCWU1299
Supplier:  Palbam