ContactUsLinkComponent
Diffraction Tube teltron electron S, max. filament voltage: 6.3 V AC, max. anode voltage: 5 kV, Anode current: approx. 0.1 mA at 4 k V, Lattice constant of graphite: d10 = 0.213 nm d11 = 0.123 nm. 1 * 1 items
Catalog # BINH1013889
Supplier:  3B Scientific
Diffraction Tube teltron electron S, max. filament voltage: 6.3 V AC, max. anode voltage: 5 kV, Anode current: approx. 0.1 mA at 4 k V, Lattice constant of graphite: d10 = 0.213 nm d11 = 0.123 nm. 1 * 1 items
Catalog # BINH1013889
Supplier:  3B Scientific
Frequently Bought Together