PCR Workstation. CE, Single UV light and single door closure. Dimensions are 61cm x 61cm x 122cm. Designed to protect against contam iniation in sensitive PCR amplification reations. UV irradiation of working area prior to use blocks replication of N 1 * 1 items
Catalog # 135-1993
Supplier: Expedeon
Frequently Bought Together