ContactUsLinkComponent
Half-tubing LDPE Antistatic Pink 360mm, Width 360mm (opens to 720mm), Length 250m, Thickness 50µm, Cleanliness level 150 per NASA JPAG 5322.1 , Compatible for use in cleanrooms 1 * 1 Roll
Catalog # KNFCLC202.14.208.2
Supplier:  KNF CLEANROOM
Half-tubing LDPE Antistatic Pink 360mm, Width 360mm (opens to 720mm), Length 250m, Thickness 50µm, Cleanliness level 150 per NASA JPAG 5322.1 , Compatible for use in cleanrooms 1 * 1 Roll
Catalog # KNFCLC202.14.208.2
Supplier:  KNF CLEANROOM
Frequently Bought Together