Coloured cap inserts
Red
Artikelnr. 525-0688
Anbieter: Brand
Jump to:
Product Details & Documents
PP.
- For quick identification
- For microtubes 211-3201 to 211-3205 and 211-3211 to 211-3215
- Applications: −196 to +121 °C
Specifications
- Colour:Red
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



