Coloured cap inserts
Blue
Artikelnr. 525-0687
Anbieter: Brand
Jump to:
Product Details & Documents
PP.
- For quick identification
- For microtubes 211-3201 to 211-3205 and 211-3211 to 211-3215
- Applications: −196 to +121 °C
Specifications
- Colour:Blue
- Seq:01



