ContactUsLinkComponent
Blautouch - Interactive worktop 1200x750x900, Frame O , Tempered Glass 8mm (Black), Steel with powder coating epoxy-polyester (Whit e) + Windows +Scanner for bar code reading 2D 1 * 1 items
Catalog # LBRLAZ1AAADABEA0AA
Supplier:  LABORIAL
Blautouch - Interactive worktop 1200x750x900, Frame O , Tempered Glass 8mm (Black), Steel with powder coating epoxy-polyester (Whit e) + Windows +Scanner for bar code reading 2D 1 * 1 items
Catalog # LBRLAZ1AAADABEA0AA
Supplier:  LABORIAL
Frequently Bought Together