ContactUsLinkComponent
PMP2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 132 amino acids and having a mo lecular mass of 19.41kDa. PMP2 is fused to His tag at N-terminus and purified by standard chromatography tec 1 * 5 µG
Catalog # BIRBORB81280-5
Supplier:  Biorbyt
PMP2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 132 amino acids and having a mo lecular mass of 19.41kDa. PMP2 is fused to His tag at N-terminus and purified by standard chromatography tec 1 * 5 µG
Catalog # BIRBORB81280-5
Supplier:  Biorbyt