ContactUsLinkComponent
alpha Synuclein Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain of 140 amino acids having a mol ecular mass of 14.4kDa. The Recombinant Human alpha Synuclein is purified by proprietary chromatographic tec 1 * 20 µG
Catalog # BIRBORB80933-20
Supplier:  Biorbyt
alpha Synuclein Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain of 140 amino acids having a mol ecular mass of 14.4kDa. The Recombinant Human alpha Synuclein is purified by proprietary chromatographic tec 1 * 20 µG
Catalog # BIRBORB80933-20
Supplier:  Biorbyt