ContactUsLinkComponent
FABP6 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 128 amino acids and having a m olecular mass of 18 kDa. FABP6 is fused to His tag at N-terminus and purified by standard chromatography tec 1 * 25 µG
Catalog # BIRBORB81105-25
Supplier:  Biorbyt
FABP6 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 128 amino acids and having a m olecular mass of 18 kDa. FABP6 is fused to His tag at N-terminus and purified by standard chromatography tec 1 * 25 µG
Catalog # BIRBORB81105-25
Supplier:  Biorbyt