ContactUsLinkComponent
FABP4 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 132 amino acids and having a m olecular mass of 18 kDa. FABP4 is fused to His tag at N-terminus and purified by standard chromatography tec 1 * 25 µG
Catalog # BIRBORB81101-25
Supplier:  Biorbyt
FABP4 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 132 amino acids and having a m olecular mass of 18 kDa. FABP4 is fused to His tag at N-terminus and purified by standard chromatography tec 1 * 25 µG
Catalog # BIRBORB81101-25
Supplier:  Biorbyt