ContactUsLinkComponent
I-TAC (Interferon-inducible T-cell alpha chemoattractant) Human Recombinant produced in E.coli is a single, non-glycosylated, polype ptide chain containing amino acids 63-87 and having total molecular mass of kDa which includes 4 kDa His tag 1 * 5 µG
Catalog # BIRBORB80670-5
Supplier:  Biorbyt
I-TAC (Interferon-inducible T-cell alpha chemoattractant) Human Recombinant produced in E.coli is a single, non-glycosylated, polype ptide chain containing amino acids 63-87 and having total molecular mass of kDa which includes 4 kDa His tag 1 * 5 µG
Catalog # BIRBORB80670-5
Supplier:  Biorbyt