ContactUsLinkComponent
Insulin Human Recombinant produced in Yeast is two chain, glycosylated polypeptide chain containing 51 amino acids and having a mole cular mass of 5807 Dalton. Zinc content was found to be 0.4%. Insulin is purified by proprietary chromatogra 1 * 2 mg
Catalog # BIRBORB80482-2
Supplier:  Biorbyt
Insulin Human Recombinant produced in Yeast is two chain, glycosylated polypeptide chain containing 51 amino acids and having a mole cular mass of 5807 Dalton. Zinc content was found to be 0.4%. Insulin is purified by proprietary chromatogra 1 * 2 mg
Catalog # BIRBORB80482-2
Supplier:  Biorbyt