Orders LinkContactUsLinkComponent
VWR®, N-Plan Objectives
VWR®, N-Plan Objectives
N-plan objective
Catalog # 630-2005
Supplier:   VWR Collection
VWR®, N-Plan Objectives
Catalog # 630-2005
Supplier:  VWR Collection
VWR®, N-Plan Objectives
VWR®, N-Plan Objectives
N-plan objective
Price
43,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
Specifications
  • Magnification:
    10×/0,25 mm
  • Description:
    N-plan objective
  • Seq:
    208