ANION STD -PHOSPHATE(AS P)1000PP 1 * 100 ml
Catalog # ACCUICPO4P10X1
Supplier: AccuStandard
Frequently Bought Together
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
VWR®, Total Acid Number (TAN) standards
Supplier: VWR Chemicals
Taurine, Reference Standard
Supplier: USP
Trace Metals 1 in Seawater - Whole Volume, (CRM), Supelco®
Supplier: Merck
Buffers, concentrated solutions, Fluka™
Supplier: Honeywell Chemicals
5,5-(DIPHENYL-D10) HYDANTOIN 1 * 5 mg
Supplier: LGC Standards PROMOCHEM
Buffer solutions
Supplier: REAGECON
Supelco® Organic Reference Standard, Eucalyptol
Supplier: Merck
Alcohol calibration standards, Ethanol, (CRM), Supelco®
Supplier: Merck
(RS)-2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]acetic Acid Dihydrochloride 1 * 25 mg
Supplier: LGC Standards PROMOCHEM
Fonofos certified 1 * 0,1 g
Supplier: EHRENSTORFER
Ofurace certified 1 * 0,1 g
Supplier: EHRENSTORFER
Organic reference standard, Diethylnitrosoamine 100 µg/ml in methanol, Dr Ehrenstorfer
Supplier: EHRENSTORFER
ISE standards
Supplier: Hach