Orders LinkContactUsLinkComponent
Micro protein quantification set, QuantiMate
Micro protein quantification set, QuantiMate
Quantification set, 8,5 mm, Quartz
Catalog # 730-1453
Supplier:   BIOCHROM
Micro protein quantification set, QuantiMate
Catalog # 730-1453
Supplier:  BIOCHROM
Micro protein quantification set, QuantiMate
Micro protein quantification set, QuantiMate
Quantification set, 8,5 mm, Quartz
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
QuantiMate is designed for concentration and purity determinations of nucleic acids and proteins.
  • Measurement performance giving accurate and reproducible concentration results
  • Samples measured without time consuming dilutions
  • Only one cuvette is required
Dimensions are equivalent to standard cuvettes in order to work in most spectrophotometers.
Specifications
  • Window material:
    Quartz
  • Description:
    Quantification set
  • Height:
    8,5 mm
  • Seq:
    001