Order Entry
Austria
Orders LinkContactUsLinkComponent
Micro protein quantification set, QuantiMate

Micro protein quantification set, QuantiMate

QuantiMate 200, 15 mm, 0,8 µl, 200 µm, Quartz
Catalog # 730-1451
Supplier:   BIOCHROM
Micro protein quantification set, QuantiMate
Catalog # 730-1451
Supplier:  BIOCHROM
Micro protein quantification set, QuantiMate
Micro protein quantification set, QuantiMate

QuantiMate 200, 15 mm, 0,8 µl, 200 µm, Quartz

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

QuantiMate is designed for concentration and purity determinations of nucleic acids and proteins.

  • Measurement performance giving accurate and reproducible concentration results
  • Samples measured without time consuming dilutions
  • Only one cuvette is required

Dimensions are equivalent to standard cuvettes in order to work in most spectrophotometers.

Specifications
  • Path length:
    200 µm
  • Volume:
    0,8 µl
  • Window material:
    Quartz
  • Description:
    QuantiMate 200
  • Height:
    15 mm
  • Seq:
    005
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.