ContactUsLinkComponent
Demountable torch, icp-ms inert kit 1/pk. demountable torch for the inert pfa sample intro kit. recommended for use with organic sol vents. excludes injector. suits 75/77/88 series icp-ms 1 * 1 items
  HEWLG4912-80013
 :  Agilent
undefined
Demountable torch, icp-ms inert kit 1/pk. demountable torch for the inert pfa sample intro kit. recommended for use with organic sol vents. excludes injector. suits 75/77/88 series icp-ms 1 * 1 items
  HEWLG4912-80013
 :  Agilent
Frequently Bought Together