ContactUsLinkComponent
Torch one piece, ball joint ax 2.3mm inj. 1/pk. one piece quartz torch for all glass sample intro kit for axial icp. fitted with bal l joint connection to torch and 2.3 mm id injector 1 * 1 items
Catalog # VARI7910050700
Supplier:  VARIAN
Torch one piece, ball joint ax 2.3mm inj. 1/pk. one piece quartz torch for all glass sample intro kit for axial icp. fitted with bal l joint connection to torch and 2.3 mm id injector 1 * 1 items
Catalog # VARI7910050700
Supplier:  VARIAN
Frequently Bought Together