ContactUsLinkComponent
All-glass sample system, axial all glass sample intro system for axial icp. includes axial torch with ball joint, glass transfer tub e, double pass glass cyclonic spraychamber + adj. mount 1 * 1 items
Catalog # VARI9910117800
Supplier:  VARIAN
All-glass sample system, axial all glass sample intro system for axial icp. includes axial torch with ball joint, glass transfer tub e, double pass glass cyclonic spraychamber + adj. mount 1 * 1 items
Catalog # VARI9910117800
Supplier:  VARIAN
Frequently Bought Together